Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-6-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Fluoro-2-nitrobenzoic acid

H63013-22 H63013-06 H63013-14
AAH63013-06EA 52.49 USD InStock
AAH63013-06 AAH63013-14 AAH63013-22
2-Fluoro-6-nitrobenzoic acid 98%
2-Fluoro-6-nitrobenzoic acid


Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862079
CAS Number: 385-02-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty