Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-N-methyl-D-phenylalanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acidFmoc-D-N-Me-Phe-OH

H63055-03 H63055-MD H63055-06
AAH63055-MDEA 51.59 USD InStock
AAH63055-MD AAH63055-06 AAH63055-03
N-Fmoc-N-methyl-D-phenylalanine 98%
N-Fmoc-N-methyl-D-phenylalanine


Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153392
CAS Number: 138775-05-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty