Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Boc-L-phenylalanine Methyl EsterBoc-Phe-OMe

H63078-22 H63078-14 H63078-06
AAH63078-14EA 114 USD InStock
AAH63078-14 AAH63078-06 AAH63078-22
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester 95%
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester


Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Melting Pt: 36…40 °C
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00076977
CAS Number: 51987-73-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty