Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorophenethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(4-Chlorophenyl)ethylamine

H63081-14 H63081-22 H63081-06
AAH63081-22EA 327.99 USD InStock
AAH63081-22 AAH63081-14 AAH63081-06
4-Chlorophenethylamine 97%
4-Chlorophenethylamine


Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 60…65 °C/0.1 mm
Density: 1.120
Flash Pt: 106 °C(223 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty