Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Methyl-L-tyrosine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methoxy-L-phenylalanine(S)-2-Amino-3-(4-methoxyphenyl)propionic AcidH-Tyr(Me)-OH

H63096-06 H63096-03
AAH63096-03EA 33.33 USD InStock
AAH63096-03 AAH63096-06
O-Methyl-L-tyrosine 98%
O-Methyl-L-tyrosine


Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 259…261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002604
CAS Number: 6230-11-1
EINECS: 228-333-9
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty