Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

z-Tyr-OMe 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Z-Tyr-OMeN-Benzyloxycarbonyl-L-tyrosine methyl ester

H63113-22 H63113-14 H63113-06
AAH63113-22EA 720.99 USD InStock
AAH63113-22 AAH63113-14 AAH63113-06
z-Tyr-OMe 98%
z-Tyr-OMe


Formula: C₁₈H₁₉NO₅
MW: 329.352 g/mol
Melting Pt: 93…97 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038784
CAS Number: 13512-31-7
EINECS: 426-320-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty