Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Aib-OH 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Benzyloxycarbonyl-alpha-methylalanine2-(Benzyloxycarbonylamino)isobutyric acid2-(Cbz-amino)isobutyric acid2-(Carbobenzoxyamino)isobutyric acidN-Cbz-alpha-methylalanineZ-Aib-OHN-Benzyloxycarbonyl-2-methylalanine

H63119-06 H63119-03
AAH63119-06EA 123.99 USD InStock
AAH63119-06 AAH63119-03
Z-Aib-OH 98%
Z-Aib-OH


Formula: C₁₂H₁₅NO₄
MW: 237.255 g/mol
Melting Pt: 65…69 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004190
CAS Number: 15030-72-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty