Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Glutamic acid 5-ethyl ester 95%

Supplier: Thermo Scientific Chemicals
Synonyms: L-Glutamic acid 5-Ethyl ester5-Ethyl L-GlutamateH-Glu(OEt)-OH

H63168-06 H63168-14
AAH63168-06EA 35.6 USD InStock
AAH63168-06 AAH63168-14
L-Glutamic acid 5-ethyl ester 95%
L-Glutamic acid 5-ethyl ester


Formula: C₇H₁₃NO₄
MW: 175.2 g/mol
Melting Pt: 179 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037217
CAS Number: 1119-33-1
EINECS: 214-274-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty