Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-guanidine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Aminoiminomethyl)carbamic acid tert-butyl estertert-Butoxycarbonylguanidine

H63206-14 H63206-03 H63206-06
AAH63206-06EA 68.99 USD InStock
AAH63206-06 AAH63206-14 AAH63206-03
N-Boc-guanidine 95%
N-Boc-guanidine
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.


Formula: C₆H₁₃N₃O₂
MW: 159.189 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20547465
CAS Number: 219511-71-4
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty