Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methyl-L-aspartate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: L-Aspartic Acid 1-Methyl EsterH-Asp-OMe

H63221-03 H63221-MD H63221-06
AAH63221-MDEA 15 USD InStock
AAH63221-MD AAH63221-03 AAH63221-06
1-Methyl-L-aspartate 98%
1-Methyl-L-aspartate


Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 186 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00080289
CAS Number: 17812-32-7
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty