Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-(1H-indol-3-yl)propanamide 98%

Supplier: Thermo Scientific Chemicals
Synonyms: DL-Tryptophanamide

H63227-03 H63227-06 H63227-MD
AAH63227-06EA 531.69 USD InStock
AAH63227-06 AAH63227-MD AAH63227-03
2-Amino-3-(1H-indol-3-yl)propanamide 98%
2-Amino-3-(1H-indol-3-yl)propanamide


Formula: C₁₁H₁₃N₃O
MW: 203.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03826622
CAS Number: 6720-02-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty