Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Fluoro-6-nitrophenyl)morpholine 97%

Supplier: Thermo Scientific Chemicals
Warning

H63272-MD H63272-03 H63272-06
AAH63272-MDEA 23.23 USD InStock
AAH63272-MD AAH63272-03 AAH63272-06
4-(2-Fluoro-6-nitrophenyl)morpholine 97%
4-(2-Fluoro-6-nitrophenyl)morpholine


Formula: C₁₀H₁₁FN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD13689260
CAS Number: 1233955-40-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty