Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

h-d-Tyr-otbu 98%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Tyrosine tert-butyl esterH-D-Tyr-OtBu

H63274-06 H63274-03
AAH63274-03EA 33.69 USD InStock
AAH63274-03 AAH63274-06
h-d-Tyr-otbu 98%
h-d-Tyr-otbu


Formula: C₁₃H₁₉NO₃
MW: 237.299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191035
CAS Number: 87553-74-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty