Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-4-iodo-D-phenylalanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-D-Phe(4-I)-OH

H63292-03 H63292-06 H63292-MD
AAH63292-03EA 63.79 USD InStock
AAH63292-03 AAH63292-06 AAH63292-MD
N-Fmoc-4-iodo-D-phenylalanine 98%
N-Fmoc-4-iodo-D-phenylalanine


Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672557
CAS Number: 205526-29-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty