Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Thienyl)-L-alanine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-β-(2-Thienyl)-Ala-OH

H63316-06 H63316-MD H63316-03
AAH63316-06EA 668.99 USD InStock
AAH63316-06 AAH63316-MD AAH63316-03
3-(2-Thienyl)-L-alanine 95%
3-(2-Thienyl)-L-alanine


Formula: C₇H₉NO₂S
MW: 171.214 g/mol
Melting Pt: 255…263 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064335
CAS Number: 22951-96-8
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty