Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-3-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

H63317-14 H63317-22
AAH63317-14EA 82.19 USD InStock
AAH63317-14 AAH63317-22
4-Fluoro-3-nitrotoluene 98%
4-Fluoro-3-nitrotoluene


Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 241° dec (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007060
CAS Number: 446-11-7
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty