Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-[2-(Boc-amino)ethyl]aniline 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl 4-aminophenethylcarbamate4-(2-Aminoethyl)aniline, 4-BOC protected

H63324-06 H63324-03
AAH63324-06EA 413.99 USD InStock
AAH63324-06 AAH63324-03
4-[2-(Boc-amino)ethyl]aniline 97%
4-[2-(Boc-amino)ethyl]aniline


Formula: C₁₃H₂₀N₂O₂
MW: 236.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663961
CAS Number: 94838-59-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty