Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Cit-OHNalpha-Fmoc-L-citrulline

H63342-14 H63342-03 H63342-06
AAH63342-06EA 86.89 USD InStock
AAH63342-06 AAH63342-14 AAH63342-03
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid


Formula: C21H23N3O5
MW: 397.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151943
CAS Number: 133174-15-9
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty