Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-2-Cyclohexylglycine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-alpha-Aminocyclohexylacetic AcidH-D-Chg-OH

H63381-06 H63381-03 H63381-14
AAH63381-06EA 157.99 USD InStock
AAH63381-06 AAH63381-03 AAH63381-14
D-2-Cyclohexylglycine 95%
D-2-Cyclohexylglycine


Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311678
CAS Number: 14328-52-0
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty