Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-D-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-D-Phe-OH

H63382-22 H63382-06 H63382-14
AAH63382-22EA 479.99 USD InStock
AAH63382-22 AAH63382-06 AAH63382-14
N-Acetyl-D-phenylalanine 95%
N-Acetyl-D-phenylalanine


Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
Melting Pt: 165…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002664
CAS Number: 10172-89-1
EINECS: 233-447-7
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty