Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-D-Glu(OtBu)-OH 95%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Glutamic acid 5-tert-butyl ester

H63463-03 H63463-14 H63463-06
AAH63463-03EA 33.49 USD InStock
AAH63463-03 AAH63463-14 AAH63463-06
H-D-Glu(OtBu)-OH 95%
H-D-Glu(OtBu)-OH


Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153437
CAS Number: 45125-00-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty