Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitro-D-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-2-Amino-3-(3-nitrophenyl)propanoic acid

H63483-MD H63483-03
AAH63483-MDEA 114.99 USD InStock
AAH63483-MD AAH63483-03
3-Nitro-D-phenylalanine 95%
3-Nitro-D-phenylalanine


Formula: C₉H₁₀N₂O₄
MW: 210.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117829
CAS Number: 169530-97-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty