Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-2,3-Epoxy-1-propanol ≥99%, ee 99% ee

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-(-)-Glycidol(S)-Oxiran-2-ylmethanol

H63502-06 H63502-03 H63502-14
AAH63502-06EA 123.99 USD InStock
AAH63502-06 AAH63502-03 AAH63502-14
(S)-(-)-2,3-Epoxy-1-propanol ≥99%, ee 99% ee
(S)-(-)-2,3-Epoxy-1-propanol
(S)-(-)-Glycidol


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 66…67 °C/19 mm
Density: 1.116
Flash Pt: 66 °C(151 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00074874
CAS Number: 60456-23-7
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty