Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(4-Thiazolyl)-L-alanine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-2-Amino-3-(thiazol-4-yl)propanoic acid

H63511-MD H63511-03
AAH63511-MDEA 126.99 USD InStock
AAH63511-MD AAH63511-03
3-(4-Thiazolyl)-L-alanine 95%
3-(4-Thiazolyl)-L-alanine


Formula: C₆H₈N₂O₂S
MW: 172.202 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079677
CAS Number: 119433-80-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty