Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Naphthyl)-D-alanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-alpha-Amino-2-naphthalenepropionic Acid

H63532-03 H63532-06 H63532-14
AAH63532-03EA 74.59 USD InStock
AAH63532-03 AAH63532-14 AAH63532-06
3-(2-Naphthyl)-D-alanine 95%
3-(2-Naphthyl)-D-alanine


Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038546
CAS Number: 76985-09-6
EINECS: 616-416-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty