Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-amino-6-bromopyrazine-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 3-amino-6-bromopyrazine-2-carboxylate

H63537-03 H63537-MD H63537-06
AAH63537-03EA 75.18 USD InStock
AAH63537-03 AAH63537-06 AAH63537-MD
Methyl-3-amino-6-bromopyrazine-2-carboxylate 97%
Methyl-3-amino-6-bromopyrazine-2-carboxylate


Formula: C6H6BrN3O2
MW: 232.03 g/mol
Melting Pt: 172…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00834964
CAS Number: 6966-01-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty