Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-aminopyrazine-2-carboxamide 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Aminopyrazine-2-carboxamide

H63553-06 H63553-MD H63553-03
AAH63553-MDEA 77.29 USD InStock
AAH63553-MD AAH63553-06 AAH63553-03
3-aminopyrazine-2-carboxamide 96%
3-aminopyrazine-2-carboxamide


Formula: C₅H₆N₄O
MW: 138.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407446
CAS Number: 32587-10-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty