Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester4-tert-Butyl N-Boc-L-aspartateN-Boc-L-aspartic Acid 4-tert-Butyl EsterBoc-Asp(OtBu)-OH

H63554-14 H63554-06
AAH63554-06EA 54.49 USD InStock
AAH63554-06 AAH63554-14
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate


Formula: C₁₃H₂₃NO₆
MW: 289.33 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00076912
CAS Number: 1676-90-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty