Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-L-phenylalanine 97%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Phe(4-Cl)-OHL-4-Chlorophenylalanine

H63573-03 H63573-06 H63573-MD
AAH63573-06EA 680.99 USD InStock
AAH63573-06 AAH63573-03 AAH63573-MD
4-Chloro-L-phenylalanine 97%
4-Chloro-L-phenylalanine


Formula: C₉H₁₀ClNO₂
MW: 199.6 g/mol
Melting Pt: 235…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063065
CAS Number: 14173-39-8
EINECS: 238-023-5
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty