Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(benzyloxy)pyrrolidine-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Hyp(Bzl)-OH

H63582-03 H63582-06 H63582-MD
AAH63582-03EA 108.99 USD InStock
AAH63582-03 AAH63582-06 AAH63582-MD
(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(benzyloxy)pyrrolidine-2-carboxylic acid 98%
(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(benzyloxy)pyrrolidine-2-carboxylic acid


Formula: C₂₇H₂₅NO₅
MW: 443.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237023
CAS Number: 174800-02-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty