Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-1H-indole-3-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Nitroindole-3-carboxaldehyde

H63595-MD H63595-03 H63595-06
AAH63595-03EA 94.89 USD InStock
AAH63595-03 AAH63595-06 AAH63595-MD
4-Nitro-1H-indole-3-carbaldehyde 97%
4-Nitro-1H-indole-3-carbaldehyde


Formula: C₉H₆N₂O₃
MW: 190.158 g/mol
Melting Pt: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD04973979
CAS Number: 10553-11-4
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty