Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-thiourea 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyl carbamothioylcarbamate

H63599-MD H63599-03 H63599-06
AAH63599-06EA 367.99 USD InStock
AAH63599-06 AAH63599-03 AAH63599-MD
N-Boc-thiourea 97%
N-Boc-thiourea


Formula: C₆H₁₂N₂O₂S
MW: 176.234 g/mol
Melting Pt: 142 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04039987
CAS Number: 268551-65-1
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty