Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2-Amino-3-(tritylthio)propanamide 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Cys(Trt)-NH₂

H63628-03 H63628-06
AAH63628-06EA 169.4 USD InStock
AAH63628-06 AAH63628-03
(R)-2-Amino-3-(tritylthio)propanamide 98%
(R)-2-Amino-3-(tritylthio)propanamide


Formula: C22H22N2OS
MW: 362.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22126061
CAS Number: 166737-85-5
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty