Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-D-threonine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-D-threonineBoc-D-Thr-OH

H63647-14 H63647-06
AAH63647-06EA 97.09 USD InStock
AAH63647-06 AAH63647-14
N-(tert-Butoxycarbonyl)-D-threonine 95%
N-(tert-Butoxycarbonyl)-D-threonine


Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037807
CAS Number: 55674-67-4
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty