Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-tert-Butyl-1-methyl-L-glutamate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester HydrochlorideH-Glu(OtBu)-OMe.HCl

H63661-06 H63661-03
AAH63661-03EA 76.79 USD InStock
AAH63661-03 AAH63661-06
5-tert-Butyl-1-methyl-L-glutamate hydrochloride 98%
5-tert-Butyl-1-methyl-L-glutamate hydrochloride


Formula: C₁₀H₁₉NO₄
MW: 253.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153438
CAS Number: 6234-01-1
EINECS: 228-348-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty