Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D-Serine methyl ester hydrochlorideH-D-Ser-OMe·HCl

H63685-22 H63685-14 H63685-06
AAH63685-06EA 26.99 USD InStock
AAH63685-06 AAH63685-14 AAH63685-22
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride


Formula: C4H10ClNO3
MW: 155.6 g/mol
Melting Pt: 163…166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066121
CAS Number: 5874-57-7
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty