Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Boc-2-azabicyclo[2.2.1]hept-5-ene 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyl 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylate2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylic acid tert-Butyl ester2-Boc-2-azabicyclo[2.2.1]hept-5-ene

H63698-06 H63698-03 H63698-MD
AAH63698-MDEA 49.79 USD InStock
AAH63698-MD AAH63698-06 AAH63698-03
2-Boc-2-azabicyclo[2.2.1]hept-5-ene 98%
2-Boc-2-azabicyclo[2.2.1]hept-5-ene


Formula: C₁₁H₁₇NO₂
MW: 195.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22416354
CAS Number: 188345-71-3
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty