Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dichloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H63749-06 H63749-03
AAH63749-06EA 133.99 USD InStock
AAH63749-06 AAH63749-03
3,4-Dichloropyridine 98%
3,4-Dichloropyridine


Formula: C₅H₃Cl₂N
Boiling Pt: 182…183 °C
Melting Pt: 22…24 °C
Storage Temperature: Ambient
MDL Number: MFCD01861989
CAS Number: 55934-00-4
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty