Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-4,4'-Biphenylalanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-4-Phenyl-D-Phe-OH

H63785-MD H63785-03
AAH63785-MDEA 103.99 USD InStock
AAH63785-MD AAH63785-03
D-4,4'-Biphenylalanine 98%
D-4,4'-Biphenylalanine


Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318879
CAS Number: 170080-13-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty