Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodo-L-tyrosine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Tyr(3-I)-OH(S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid

H63789-06 H63789-03
AAH63789-03EA 46.34 USD InStock
AAH63789-03 AAH63789-06
3-Iodo-L-tyrosine 98%
3-Iodo-L-tyrosine


Formula: C₉H₁₀INO₃
MW: 307.1 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002608
CAS Number: 70-78-0
EINECS: 200-744-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty