Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H63825-06 H63825-03 H63825-MD
AAH63825-06EA 772.99 USD InStock
AAH63825-06 AAH63825-MD AAH63825-03
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride


Formula: C6H12ClNO
MW: 149.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800611
CAS Number: 54745-74-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty