Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodo-5-trifluoromethylpyridine 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Iodo-5-(trifluoromethyl)pyridine

H63848-MD H63848-06 H63848-03
AAH63848-03EA 99.59 USD InStock
AAH63848-03 AAH63848-MD AAH63848-06
2-Iodo-5-trifluoromethylpyridine 96%
2-Iodo-5-trifluoromethylpyridine


Formula: C6H3F3IN
MW: 272.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741351
CAS Number: 100366-75-4
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty