Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(ɑ)-Boc-L-lysine 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-Lys-OHNalpha-Boc-L-lysine

H63859-22 H63859-06 H63859-14
AAH63859-14EA 252.99 USD InStock
AAH63859-14 AAH63859-06 AAH63859-22
N-(ɑ)-Boc-L-lysine 97%
N-(alpha)-Boc-L-lysine


Formula: C11H22N2O4
MW: 246.3 g/mol
Melting Pt: ca 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038203
CAS Number: 13734-28-6
EINECS: 237-303-4
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty