Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-6-iodopyridazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H63879-06 H63879-03
AAH63879-06EA 271.99 USD InStock
AAH63879-06 AAH63879-03
3-Chloro-6-iodopyridazine 97%
3-Chloro-6-iodopyridazine


Formula: C₄H₂ClIN₂
MW: 240.43 g/mol
Melting Pt: 110…120 °C
Storage Temperature: Ambient
MDL Number: MFCD08275187
CAS Number: 135034-10-5
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty