Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Tyr(BZL)-OMe·HCl 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Tyr(Bzl)-OMe·HCl

H63908-03 H63908-06
AAH63908-06EA 115.99 USD InStock
AAH63908-06 AAH63908-03
H-Tyr(BZL)-OMe·HCl 98%
H-Tyr(BZL)-OMe·HCl


Formula: C₁₇H₂₀ClNO₃
MW: 321.801 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080888
CAS Number: 34805-17-9
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty