Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Cha-OHN-Fmoc-3-cyclohexyl-L-alanine

H63910-03 H63910-06
AAH63910-06EA 188.99 USD InStock
AAH63910-06 AAH63910-03
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine


Formula: C₂₄H₂₇NO₄
MW: 393.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065614
CAS Number: 135673-97-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty