Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-β-Aminophenethyl alcohol 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DL-2-Phenylglycinol2-Amino-2-phenylethanolDL-Phenylglycinol

H63932-14 H63932-06 H63932-22
AAH63932-14EA 113.99 USD InStock
AAH63932-14 AAH63932-22 AAH63932-06
(±)-β-Aminophenethyl alcohol 95%
(±)-β-Aminophenethyl alcohol


Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00130145
CAS Number: 7568-92-5
UN: 3263
ADR: "8",III
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty