Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(3-Pyridyl)-L-alanine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-β-(3-Pyridyl)-Ala-OH

H63941-06 H63941-03 H63941-MD
AAH63941-03EA 178.99 USD InStock
AAH63941-03 AAH63941-MD AAH63941-06
3-(3-Pyridyl)-L-alanine 98%
3-(3-Pyridyl)-L-alanine


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 248…252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079676
CAS Number: 64090-98-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty