Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-bromo-5-methylthiazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-5-methylthiazole

H63951-06 H63951-03
AAH63951-03EA 59.79 USD InStock
AAH63951-03 AAH63951-06
2-bromo-5-methylthiazole 98%
2-bromo-5-methylthiazole


Formula: C₄H₄BrNS
MW: 178.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460610
CAS Number: 41731-23-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty