Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Glu-OtBu 95%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-tert-Butyl N-Benzyloxycarbonyl-L-glutamateN-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl esterN-Benzyloxycarbonyl-L-glutamic acid 1-tert-Butyl esterN-Cbz-L-glutamic acid 1-tert-Butyl esterN-Carbobenzoxy-L-glutamic acid 1-tert-Butyl ester1-tert-Butyl N-Cbz-L-glutamateZ-Glu-OtBu1-tert-Butyl N-Carbobenzoxy-L-glutamate

H63954-03 H63954-MD H63954-06
AAH63954-03EA 78.79 USD InStock
AAH63954-03 AAH63954-MD AAH63954-06
Z-Glu-OtBu 95%
Z-Glu-OtBu


Formula: C₁₇H₂₃NO₆
MW: 337.372 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077016
CAS Number: 5891-45-2
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty